missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | MRPS12 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:20 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18437931
|
Novus Biologicals
NBP2-13620-25ul |
25ul |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18105690
|
Novus Biologicals
NBP2-13620 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MRPS12 Polyclonal specifically detects MRPS12 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MRPS12 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 6183 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: MSWSGLLHGLNTSLTCGPALVPRLWATCSMATLNQMHRL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:20 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 28S ribosomal protein S12, mitochondrial, mitochondrial ribosomal protein S12, MPR-S12, MRP-S12, MT-RPS12, ribosomal protein, mitochondrial, S12, RPMS12, RPS12, RPSM12S12mt | |
| MRPS12 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title