missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MRPS12 Polyclonal antibody specifically detects MRPS12 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
Specifications
| Antigen | MRPS12 |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 405 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 28S ribosomal protein S12, mitochondrial, mitochondrial ribosomal protein S12, MPR-S12, MRP-S12, MT-RPS12, ribosomal protein, mitochondrial, S12, RPMS12, RPS12, RPSM12S12mt |
| Host Species | Rabbit |
| Immunogen | mitochondrial ribosomal protein S12. (Uniprot# O15235) |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?