missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS12 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93202-0.1ml
This item is not returnable.
View return policy
Description
MRPS12 Polyclonal antibody specifically detects MRPS12 in Human samples. It is validated for Western Blot
Specifications
| MRPS12 | |
| Polyclonal | |
| Western Blot 1:1000-1:2000 | |
| 28S ribosomal protein S12, mitochondrial, mitochondrial ribosomal protein S12, MPR-S12, MRP-S12, MT-RPS12, ribosomal protein, mitochondrial, S12, RPMS12, RPS12, RPSM12S12mt | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 30-138 of human MRPS12 (NP_203527.1). MATLNQMHRLGPPKRPPRKLGPTEGRPQLKGVVLCTFTRKPKKPNSANRKCCRVRLSTGREAVCFIPGEGHTLQEHQIVLVEGGRTQDLPGVKLTVVRGKYDCGHVQKK | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6183 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction