missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPS11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560497
This item is not returnable.
View return policy
Description
Immunogen sequence: GFRNAKKGTG IAAQTAGIAA AARAKQKGVI HIRVVVKGLG PGRLSAMHGL IMGGLEVISI TDNTPIPHNG Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that contains a high level of sequence similarity with ribosomal protein S11P family members. A pseudogene corresponding to this gene is found on chromosome 20. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| MRPS11 | |
| Polyclonal | |
| Unconjugated | |
| MRPS11 | |
| 0710005I03Rik; 28S ribosomal protein S11, mitochondrial; C79873; Cervical cancer proto-oncogene 2 protein; HCC2; HCC-2; mitochondrial ribosomal protein S11; Mitochondrial small ribosomal subunit protein uS11m; MRPS11; MRP-S11; RGD1559901; RPMS11; S11mt | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64963 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P82912 | |
| MRPS11 | |
| Recombinant protein corresponding to Human MRPS11. Recombinant protein control fragment (Product #RP-100639). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction