missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20959-1-AP-20UL
This item is not returnable.
View return policy
Description
KIAA0391, also known as MRPP3 (mitochondrial ribonuclease P protein 3), is a 583 amino acid protein that is located in the mitochondria. KIAA0391 functions in tRNA maturation and is a component of mitochondrial ribonuclease P which cleaves tRNA at the 5′-end. Mitochondrial ribonuclease P is an enzyme composed of RG9MTD1, MRPP22 and KIAA0391 subunits. Due to alternative splicing events, KIAA0391 exists as four isoforms. The gene encoding KIAA0391 maps to human chromosome 14, which contains about 700 genes and makes up about 3.5% of human cellular DNA.Specifications
| MRPP3 | |
| Polyclonal | |
| Unconjugated | |
| KIAA0391 | |
| KIAA0391, MRPP3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 299050, 66132, 9692 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| B5DF07, O15091, Q8JZY4 | |
| Kiaa0391, Mrpp3 | |
| MRPP3 Fusion Protein Ag15196 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction