missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MRPL9 Polyclonal antibody specifically detects MRPL9 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
Specifications
| Antigen | MRPL9 |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | PE |
| Formulation | PBS |
| Gene Alias | L9mt, mitochondrial ribosomal protein L9, MRP-L9,39S ribosomal protein L9, mitochondrial |
| Host Species | Rabbit |
| Immunogen | mitochondrial ribosomal protein L9. (Uniprot# Q9BYD2) |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?