missing translation for 'onlineSavingsMsg'
Learn More

MRPL54 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£149.00 - £366.00

Specifications

Antigen MRPL54
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18671512
View Documents
Novus Biologicals
NBP2-95150-0.02ml
0.02 mL
£149.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18604902
View Documents
Novus Biologicals
NBP2-95150-0.1ml
0.1 mL
£366.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MRPL54 Polyclonal antibody specifically detects MRPL54 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

MRPL54
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
L54mt, mitochondrial ribosomal protein L54, MRP-L54,39S ribosomal protein L54, mitochondrial
Recombinant fusion protein containing a sequence corresponding to amino acids 1-138 of human MRPL54 (NP_758455.1). MATKRLFGATRTWAGWGAWELLNPATSGRLLARDYAKKPVMKGAKSGKGAVTSEALKDPDVCTDPVQLTTYAMGVNIYKEGQDVPLKPDAEYPEWLFEMNLGPPKTLEELDPESREYWRRLRKQNIWRHNRLSKNKRL
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin
Polyclonal
Purified
RUO
PBS (pH 7.3), 50% glycerol
116541
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.