missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL50 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£342.00 - £470.00
Specifications
| Antigen | MRPL50 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18486210
|
Novus Biologicals
NBP1-83651-25ul |
25 μL |
£342.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18203407
|
Novus Biologicals
NBP1-83651 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MRPL50 Polyclonal specifically detects MRPL50 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| MRPL50 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 39S ribosomal protein L50, mitochondrial, FLJ20493, L50mt, mitochondrial 39S ribosomal protein L50, mitochondrial ribosomal protein L50, MRP-L50FLJ21990 | |
| MRPL50 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 54534 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:MAERSVSGITRRVFMWTVSGTPCREFWSRFRKEKEPVVVETVEEKKEPILVCPPLRSRAYTPPEDLQSRLESYVKEV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title