missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL46 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94483-0.1ml
This item is not returnable.
View return policy
Description
MRPL46 Polyclonal antibody specifically detects MRPL46 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MRPL46 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| C15orf4, L46mt, LIECG2chromosome 15 open reading frame 4, MGC22762, mitochondrial ribosomal protein L46, MRP-L46, P2ECSL39S ribosomal protein L46, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-283 of mouse MRPL46 (NP_075820.1). MAAPVGRTLLGLARGWRQLDRFWAGSSRGLSLEAASSSSRSPWRLSGALCLQRPPLITKALTPLQEEMAGLLQQIEVERSLYSDHELRALDEAQRLAKKKADLYDEEQEQGITLAQDLEDMWEQAFLQFRPGARETEADKKNDRTSLHRKLDRNLVLLVREKLGDQDVWMLPQVEWQPGETLRGTAERILATLSENNMEAKFLGNAPCGHYKFKFPKAIQTESDLGVKVFFFKALLLTGDFVQAGKKSRHVWASKEELGDYLQPKYLAQVRRFLLDSDGLSCL | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 26589 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction