missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MRPL46 Polyclonal antibody specifically detects MRPL46 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | MRPL46 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | C15orf4, L46mt, LIECG2chromosome 15 open reading frame 4, MGC22762, mitochondrial ribosomal protein L46, MRP-L46, P2ECSL39S ribosomal protein L46, mitochondrial |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-283 of mouse MRPL46 (NP_075820.1). MAAPVGRTLLGLARGWRQLDRFWAGSSRGLSLEAASSSSRSPWRLSGALCLQRPPLITKALTPLQEEMAGLLQQIEVERSLYSDHELRALDEAQRLAKKKADLYDEEQEQGITLAQDLEDMWEQAFLQFRPGARETEADKKNDRTSLHRKLDRNLVLLVREKLGDQDVWMLPQVEWQPGETLRGTAERILATLSENNMEAKFLGNAPCGHYKFKFPKAIQTESDLGVKVFFFKALLLTGDFVQAGKKSRHVWASKEELGDYLQPKYLAQVRRFLLDSDGLSCL |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?