missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL44 Monoclonal Antibody (OTI3H5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527323
This item is not returnable.
View return policy
Description
MRPL44 Monoclonal Antibody for Western Blot, IHC (P)
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein.
Specifications
| MRPL44 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H9J2 | |
| MRPL44 | |
| Full length human recombinant protein of MRPL44 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3H5 | |
| Unconjugated | |
| MRPL44 | |
| 1810030E18Rik; 39S ribosomal protein L44, mitochondrial; 5730593H20Rik; AI463302; COXPD16; l44; L44mt; Mitochondrial large ribosomal subunit protein mL44; mitochondrial ribosomal protein L44; MRPL44; MRP-L44; RM44 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 65080 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction