missing translation for 'onlineSavingsMsg'
Learn More

MRPL44 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18416529 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416529 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416529 Supplier Novus Biologicals Supplier No. NBP296998MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MRPL44 Polyclonal antibody specifically detects MRPL44 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL44
Applications Western Blot, ELISA, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias 39S ribosomal protein L44, mitochondrial, EC 3.1.26, EC 3.1.26.-, FLJ12701, FLJ13990, FLJ37688, L44MT, mitochondrial ribosomal protein L44, MRP-L44
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human MRPL44 (Uniprot#: Q9H9J2; Met1-Ser332)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Endocrinology
Primary or Secondary Primary
Gene ID (Entrez) 65080
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.