missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL43 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570588
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 79%; Guinea Pig: 79%; Horse: 79%; Human: 100%; Mouse: 79%; Rat: 83%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. This gene and the gene for a semaphorin class 4 protein (SEMA4G) overlap at map location 10q24.31 and are transcribed in opposite directions. Sequence analysis identified multiple transcript variants encoding at least four different protein isoforms.
Specifications
| MRPL43 | |
| Polyclonal | |
| Unconjugated | |
| MRPL43 | |
| 39S ribosomal protein L43, mitochondrial; 4930442D21Rik; bMRP36a; L43mt; Mitochondrial large ribosomal subunit protein mL43; Mitochondrial ribosomal protein bMRP36a; mitochondrial ribosomal protein L43; MRPL43; MRP-L43; MRPL43 protein; putative; retina-specific 15.7 kDa protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 84545 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N983 | |
| MRPL43 | |
| synthetic peptide directed towards the middle region of human MRPL43 (aa 7-56). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction