missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL40 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00064976-B01P
This item is not returnable.
View return policy
Description
MRPL40 Polyclonal antibody specifically detects MRPL40 in Human samples. It is validated for Western Blot
Specifications
| MRPL40 | |
| Polyclonal | |
| Western Blot 1:500 | |
| AAH09707 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| FLJ41774, L40mt, MGC9400, mitochondrial ribosomal protein L40, MRP-L22,39S ribosomal protein L40, mitochondrial, MRP-L40, NLVCFURIM, nuclear localization signal containing protein deleted in velocardiofacialsyndrome, nuclear localization signal deleted in velocardiofacial syndrome, Nuclear localization signal-containing protein deleted in velocardiofacialsyndrome, Up-regulated in metastasis | |
| MRPL40 (AAH09707, 1 a.a. - 206 a.a.) full-length human protein. MTASVLRSISLALRPTSGLLGTWQTQLRETHQRASLLSFWELIPMRSEPLRKKKKVDPKKDQEAKERLKRKIRKLEKATQELIPIEDFITPLKFLDKARERPQVELTFEETERRALLLKKWSLYKQQERKMERDTIRAMLEAQQEALEELQLESPKLHAEAIKRDPNLFPFEKEGPHYTPPIPNYQPPEGRYNDITKVYTQVEFKR | |
| 0.05 mg | |
| Endocrinology | |
| 64976 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction