missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL37 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115591
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MRPL37.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit.
Specifications
| MRPL37 | |
| Polyclonal | |
| Unconjugated | |
| MRPL37 | |
| 2300004O14Rik; 39S ribosomal protein L2, mitochondrial; 39S ribosomal protein L37, mitochondrial; AI132596; HSPC235; L2mt; L37mt; MGC878; Mitochondrial large ribosomal subunit protein mL37; mitochondrial ribosomal protein L37; MRPL2; MRP-L2; MRPL37; MRP-L37; ribosomal protein, mitochondrial, L2; RPML2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51253 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q9BZE1 | |
| MRPL37 | |
| A synthesized peptide derived from human MRPL37(Accession Q9BZE1), corresponding to amino acid residues P57-F107. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction