missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL36 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100804
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total L36mt.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. MRPL36 encodes a 39S subunit protein. A pseudogene corresponding to this gene is found on chromosome 2p.
Specifications
| MRPL36 | |
| Polyclonal | |
| Unconjugated | |
| MRPL36 | |
| 39S ribosomal protein L36, mitochondrial; AI646041; BRCA1-interacting protein 1; BRIP1; L36mt; Mitochondrial large ribosomal subunit protein bL36m; mitochondrial ribosomal protein L36; MRPL36; MRP-L36; PRPL36; putative BRCA1-interacting protein; RPMJ | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64979, 94066 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q99N90, Q9P0J6 | |
| MRPL36 | |
| A synthesized peptide derived from human MRPL36(Accession Q9P0J6), corresponding to amino acid residues A47-R97. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction