missing translation for 'onlineSavingsMsg'
Learn More

MRPL36 Antibody [DyLight 488], Novus Biologicals™

Product Code. 30760983 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30760983 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30760983 Supplier Novus Biologicals Supplier No. NBP347037G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MRPL36 Polyclonal antibody specifically detects MRPL36 in Human samples. It is validated for ELISA,Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL36
Applications ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias BRCA1-interacting protein 1, BRIP1MGC104245, L36mtMRP-L36PRPL36, mitochondrial ribosomal protein L36,39S ribosomal protein L36, mitochondrial, putative BRCA1-interacting protein, RPMJ
Host Species Rabbit
Immunogen mitochondrial ribosomal protein L36. (Uniprot# Q9P0J6)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64979
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.