missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31066
This item is not returnable.
View return policy
Description
MRPL3 Polyclonal specifically detects MRPL3 in Human samples. It is validated for Western Blot.
Specifications
| MRPL3 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| MRPL3 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human MRPL3. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 11222 | |
| Human | |
| IgG |
| Western Blot | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000 | |
| 39S ribosomal protein L3, mitochondrial, mitochondrial ribosomal protein L3, MRL3mitochondrial, L3, RPML3 | |
| Rabbit | |
| 39 kDa | |
| 100 μL | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction