missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL28 Rabbit anti-Human, Mouse, Rat, Clone: 3B9F2, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | MRPL28 |
|---|---|
| Clone | 3B9F2 |
| Dilution | Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 1:50 - 1:200 |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18306672
|
Novus Biologicals
NBP3-15267-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18399903
|
Novus Biologicals
NBP3-15267-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MRPL28 Monoclonal antibody specifically detects MRPL28 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
| MRPL28 | |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 1:50 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| L28mt, MAAT139S ribosomal protein L28, mitochondrial, Melanoma antigen p15, melanoma-associated antigen recognised by cytotoxic T lymphocytes, Melanoma-associated antigen recognized by T lymphocytes, MGC8499, mitochondrial ribosomal protein L28, MRP-L28, p15 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MRPL28 (Q13084). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 3B9F2 | |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| DNA Repair, DNA replication Transcription Translation and Splicing | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 10573 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title