missing translation for 'onlineSavingsMsg'
Learn More

MRPL28 Antibody, Novus Biologicals™

Product Code. 18396908 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396908 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18396908 Supplier Novus Biologicals Supplier No. H00010573B02P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

MRPL28 Polyclonal antibody specifically detects MRPL28 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL28
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_006419.2
Gene Alias L28mt, MAAT139S ribosomal protein L28, mitochondrial, Melanoma antigen p15, melanoma-associated antigen recognised by cytotoxic T lymphocytes, Melanoma-associated antigen recognized by T lymphocytes, MGC8499, mitochondrial ribosomal protein L28, MRP-L28, p15
Host Species Mouse
Immunogen MRPL28 (NP_006419.2, 1 a.a. - 256 a.a.) full-length human protein. MPLHKYPVWLWKRLQLREGICSRLPGHYLRSLEEERTPTPVHYRPHGAKFKINPKNGQRERVEDVPIPIYFPPESQRGLWGGEGWILGQIYANNDKLSKRLKKVWKPQLFEREFYSEILDKKFTVTVTMRTLDLIDEAYGLDFYILKTPKEDLCSKFGMDLKRGMLLRLARQDPQLHPEDPERRAAIYDKYKEFAIPEEEAEWVGLTLEEAIEKQRLLEEKDPVPLFKIYVAELIQQLQQQALSEPAVVQKRASGQ
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 10573
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.