missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL24 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541909
This item is not returnable.
View return policy
Description
Peptide sequence: RRRPVVVEPI SDEDWYLFCG DTVEILEGKD AGKQGKVVQV IRQRNWVVVG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein which is more than twice the size of its E. coli counterpart. Sequence analysis identified two transcript variants that encode the same protein.
Specifications
| MRPL24 | |
| Polyclonal | |
| Unconjugated | |
| mrpl24 | |
| 2010005E08Rik; 2810470K06Rik; 39S ribosomal protein L24, mitochondrial; 6720473G22Rik; AA407670; FLJ20917; hypothetical protein LOC532203; L24mt; MGC22737; MGC9831; Mitochondrial large ribosomal subunit protein uL24m; mitochondrial ribosomal protein L24; MRP-L18; MRPL24; MRP-L24; MRPL24 protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 79590 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96A35 | |
| mrpl24 | |
| Synthetic peptide directed towards the N-terminal of human MRPL24 (aa 40-89). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction