missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL23 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562184
This item is not returnable.
View return policy
Description
Immunogen sequence: KRRDHRNVRI KKPDYKVAYV QLAHGQTFTF PDLFPEKDES PEGSAADDLY SMLEEERQQR QSSDPRRGGV PSWFGL Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 74%.
MRPL23 are mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. MRPL23 encodes a 39S subunit protein and is biallelically expressed, despite its location within a region of imprinted genes on chromosome 11.
Specifications
| MRPL23 | |
| Polyclonal | |
| Unconjugated | |
| MRPL23 | |
| 39S ribosomal protein L23, mitochondrial; FLJ45387; L23 mitochondrial-related protein; L23mrp; L23mt; Mitochondrial large ribosomal subunit protein uL23m; mitochondrial ribosomal protein L23; Mrpl23; MRP-L23; Ribosomal protein L23-like; ribosomal protein L23-related product homolog; ribosomal protein related to L23 (mitochondrial); rL23MRP; Rpl23; RPL23L; Rpl23-rs | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6150 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16540 | |
| MRPL23 | |
| Recombinant protein corresponding to Human MRPL23. Recombinant protein control fragment (Product #RP-100653). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction