missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL21 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531876
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, Raji. Predicted reactivity: Mouse (84%), Rat (85%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Multiple transcript variants encoding different isoforms were identified through sequence analysis although some may be subject to nonsense-mediated decay (NMD).
Specifications
| MRPL21 | |
| Polyclonal | |
| Unconjugated | |
| MRPL21 | |
| 39S ribosomal protein L21, mitochondrial; BC028768; D9Wsu149; L21mt; Mitochondrial large ribosomal subunit protein bL21m; mitochondrial ribosomal protein L21; Mrpl21; MRP-L21; Unknown (protein for MGC:133585) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 219927 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q7Z2W9 | |
| MRPL21 | |
| Recombinant protein encompassing a sequence within the center region of human MRPL21. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction