missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL21 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19402
This item is not returnable.
View return policy
Description
MRPL21 Polyclonal specifically detects MRPL21 in Human samples. It is validated for Western Blot.
Specifications
| MRPL21 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| L21mt, MGC62013, mitochondrial ribosomal protein L21, MRP-L21,39S ribosomal protein L21, mitochondrial | |
| Rabbit | |
| 23 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Bovine (82%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q7Z2W9 | |
| MRPL21 | |
| Recombinant protein encompassing a sequence within the center region of human MRPL21. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 219927 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction