missing translation for 'onlineSavingsMsg'
Learn More

MRPL21 Antibody [CoraFluor™ 1], Novus Biologicals™

Product Code. 30763208 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30763208 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30763208 Supplier Novus Biologicals Supplier No. NBP347044CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MRPL21 Polyclonal antibody specifically detects MRPL21 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL21
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate CoraFluor 1
Formulation PBS
Gene Alias L21mt, MGC62013, mitochondrial ribosomal protein L21, MRP-L21,39S ribosomal protein L21, mitochondrial
Host Species Rabbit
Immunogen mitochondrial ribosomal protein L21. (Uniprot# Q7Z2W9)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219927
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.