missing translation for 'onlineSavingsMsg'
Learn More

MRPL20 Antibody, CoraFluor™ 1, Novus Biologicals™

Product Code. 30157746
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30157746 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30157746 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP298645CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MRPL20 Polyclonal antibody specifically detects MRPL20 in Human samples. It is validated for Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL20
Applications Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate CoraFluor 1
Formulation PBS
Gene Alias 39S ribosomal protein L20, mitochondrial, FLJ10024, L20mt, MGC4779, MGC74465, mitochondrial ribosomal protein L20, MRP-L20
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MRPL20 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55052
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.