missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566721
This item is not returnable.
View return policy
Description
Immunogen sequence: PCRSADIALVA GGSRKRWIIA TENMQAGDTI LNSNHIGRMA VAAREGDAHP LGALPVGTLI NNVESEPGRG AQYIRAAGTC GVLL Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein that belongs to the EcoL2 ribosomal protein family. A pseudogene corresponding to this gene is found on chromosome 12q. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq, Jul 2014].
Specifications
| MRPL2 | |
| Polyclonal | |
| Unconjugated | |
| MRPL2 | |
| 39S ribosomal protein L2, mitochondrial; CGI-22; L2mt; Mitochondrial large ribosomal subunit protein uL2m; mitochondrial ribosomal protein L2; MRP-L14; MRPL2; MRP-L2; ribosomal protein, mitochondrial, L14; Rpml14 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51069 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5T653 | |
| MRPL2 | |
| Recombinant Human MRPL2. Recombinant protein control fragment (Product #RP-107373). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction