missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRPL19 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17228373 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17228373 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17228373 Supplier Invitrogen™ Supplier No. PA5115590

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MRPL19.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL19
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene MRPL19
Gene Accession No. P49406, Q9D338
Gene Alias 39S ribosomal protein L15, mitochondrial; 39S ribosomal protein L19, mitochondrial; 9030416F12Rik; 9130412E02Rik; D6Ertd157e; KIAA0104; L15mt; L19mt; Mitochondrial large ribosomal subunit protein bL19m; mitochondrial ribosomal protein L19; MRPL15; MRP-L15; Mrpl19; MRP-L19; ribosomal protein, mitochondrial, L15; RLX1; RPML15
Gene Symbols MRPL19
Host Species Rabbit
Immunogen A synthesized peptide derived from human MRPL19(Accession P49406), corresponding to amino acid residues Y197-L247.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 297372, 56284, 9801
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.