missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546534
This item is not returnable.
View return policy
Description
Peptide sequence: RNPGCRFAAL STSSEPAAKP EVDPVENEAV APEFTNRNPR NLELLSVARK Sequence homology: Dog: 93%; Human: 100%; Rabbit: 79%; Rat: 92%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein that belongs to the L18P ribosomal protein family. Three polymorphic sites exist in this gene, one of which is three nt in length which causes an extra aa near the N-terminus.
Specifications
| MRPL18 | |
| Polyclonal | |
| Unconjugated | |
| Mrpl18 | |
| 1010001C05Rik; 39S ribosomal protein L18, mitochondrial; C79879; HSPC071; L18mt; Mitochondrial large ribosomal subunit protein uL18m; mitochondrial ribosomal protein L18; Mrpl18; MRP-L18 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 29074 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H0U6 | |
| Mrpl18 | |
| Synthetic peptide directed towards the N-terminal of human MRPL18 (aa 15-64). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction