missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL17 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18407R
This item is not returnable.
View return policy
Description
MRPL17 Polyclonal antibody specifically detects MRPL17 in Human samples. It is validated for Western Blot
Specifications
| MRPL17 | |
| Polyclonal | |
| Western Blot | |
| L17mt, LIP2, LYST-interacting protein 2, LYST-interacting protein LIP2, mitochondrial ribosomal protein L17, MRP-L17, MRP-L26, RPL17L, RPML26,39S ribosomal protein L17, mitochondrial | |
| Recognizes region between residues Between Between 25 and 75.. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 63875 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction