missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL15 Monoclonal Antibody (OTI7A3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526862
This item is not returnable.
View return policy
Description
MRPL15 Monoclonal Antibody for Western Blot, IHC (P)
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein that belongs to the EcoL15 ribosomal protein family. A pseudogene corresponding to this gene is found on chromosome 15q.
Specifications
| MRPL15 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9P015 | |
| Mrpl15 | |
| Human recombinant protein fragment corresponding to amino acids 28-296 of MRPL15 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7A3 | |
| Unconjugated | |
| Mrpl15 | |
| 39S ribosomal protein L15, mitochondrial; HSPC145; L15mt; Mitochondrial large ribosomal subunit protein uL15m; mitochondrial ribosomal protein L15; Mrpl15; MRP-L15; MRP-L7; ribosomal protein, mitochondrial, L7; RPML7 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 29088 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion