missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL13 Antibody (OTI6A11), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72780APC
This item is not returnable.
View return policy
Description
MRPL13 Monoclonal antibody specifically detects MRPL13 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MRPL13 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI6A11 | |
| Western Blot | |
| 39S ribosomal protein L13, mitochondrial, L13, L13A, L13mtRPML13, mitochondrial ribosomal protein L13, MRP-L13, RPL13 | |
| Full length human recombinant protein of human MRPL13 (NP_054797) produced in E.coli. | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 28998 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction