missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14795-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody reacts specifically with 21 kDa MRPL12 protein.
MRPL12 (mitochondrial ribosomal protein L12), also referred to as 5c5-2, L12mt, MRPL7 or RPML12, is a mammalian mitochondrial ribosomal protein that is involved in protein synthesis within the mitochondrion. MRPL12 is enhanced in growth-stimulated cells as a result of transcriptional activation, suggesting that it may function as a translational regulator of mitochondrial mRNAs. Impairment of MRPL12 leads to reduction in cell growth rate, decreased mitochondrial ATP production and abolition of mitochondrial oxidative phosphorylation. MRPL12 is cleaved during its translocation across the mitochondrial membrane and it exists as dimers that bind the large ribosomal subunit. MRPL12 is 198 amino acids in length, belongs to the ribosomal protein L12P family and is highly expressed in the colon.Specifications
| MRPL12 | |
| Polyclonal | |
| Unconjugated | |
| MRPL12 | |
| 5c5 2, L12mt, MRP L12, MRP L31/34, MRPL12, MRPL7, MRPL7/L12, RPML12 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 303746, 56282, 6182 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P52815, Q9DB15 | |
| Mrpl12 | |
| MRPL12 Fusion Protein Ag6504 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction