missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL12 Polyclonal Antibody

MRPL12 Polyclonal Antibody
Brand: Invitrogen PA575535
This item is not returnable.
View return policy
Description
MRPL12 Polyclonal Antibody for Western Blot
Specifications
| MRPL12 | |
| Polyclonal | |
| Unconjugated | |
| MRPL12 | |
| 0610034O11Rik; 1500031N16Rik; 39S ribosomal protein L12, mitochondrial; 5c5-2; FLJ60124; hypothetical protein LOC550391; im:6893415; L12mt; MGC8610; Mitochondrial large ribosomal subunit protein bL12m; mitochondrial ribosomal protein L12; mitochondrial ribosomal protein L7/12; MRPL12; MRP-L12; MRP-L31/34; MRPL7; MRPL7/L12; ribosomal protein, mitochondrial, L12; Rpm12; RPML12; zgc:112445 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 303746, 56282, 6182 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide; pH 7.2 | |
| P52815, Q9DB15 | |
| MRPL12 | |
| Synthetic peptide corresponding to amino acids 75-120 of Human MRPL12. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction