missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL11 Polyclonal Antibody

Description
MRPL11 Polyclonal Antibody detects endogenous levels of MRPL11 protein.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Sequence analysis identified three transcript variants that encode different isoforms. Pseudogenes corresponding to this gene are found on chromosomes 5q and 12q.
Specifications
Specifications
| Antigen | MRPL11 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | MRPL11 |
| Gene Accession No. | Q9Y3B7 |
| Gene Alias | 2410001P07Rik; 39S ribosomal protein L11, mitochondrial; CGI-113; cgi-113 protein; L11mt; mgc111024; Mitochondrial large ribosomal subunit protein uL11m; mitochondrial ribosomal protein L11; Mrpl11; MRP-L11 |
| Gene Symbols | MRPL11 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?