missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531245
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T. Predicted reactivity: Human (99%), Mouse (83%), Rat (86%), Rhesus Monkey (97%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. A pseudogene corresponding to this gene is found on chromosome 5q.
Specifications
| MRPL10 | |
| Polyclonal | |
| Unconjugated | |
| MRPL10 | |
| 0610040E02Rik; 39S ribosomal protein L10, mitochondrial; 39S ribosomal protein L8, mitochondrial; C78440; L10MT; L8mt; Mitochondrial large ribosomal subunit protein uL10m; mitochondrial ribosomal protein L10; MRPL10; MRP-L10; MRPL8; MRP-L8; Rpml8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 124995 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.68 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q7Z7H8 | |
| MRPL10 | |
| Recombinant protein encompassing a sequence within the center region of human MRPL10. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction