missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRP7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101678
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MRP7.
The MRP family is represented by nine similar ABC transporters that have the ability to transport structurally diverse lipophilic anions and operate as chemical efflux pumps. MRP7 (multidrug resistance-associated protein 7, ATPbinding cassette sub-family C member 10) is a multi-pass membrane protein that belongs to the ABC transporter family (conjugate transporter subfamily). MRP7 is involved with the ATP-dependent transport of 17β-estradiol-D-17-β-glucuronide (E217βG). MRP7 is also probably involved in cellular detoxification through its lipophilic anion extrusion capabilities. MRP7 contains two ABC transmembrane type 1 domains and two ABC transporter domains. MRP7 likely has three isoforms. Isoform 2 is the most widely expressed, while isoform 1 is predominately expressed in the spleen.
Specifications
| MRP7 | |
| Polyclonal | |
| Unconjugated | |
| ABCC10 | |
| ABCC10; ATP binding cassette subfamily C member 10; ATP-binding cassette sub-family C member 10; ATP-binding cassette, subfamily C (CFTR/MRP), member 10; ATP-binding cassette, sub-family C (CFTR/MRP), member 10; EST182763; mFLJ00002; Mrp7; Multidrug resistance-associated protein 7; SIMRP7 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 89845 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5T3U5 | |
| ABCC10 | |
| A synthesized peptide derived from human ABCC10(Accession Q5T3U5), corresponding to amino acid residues A1351-K1401. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction