missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRP6 Antibody (M6II-31), Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-42343
This item is not returnable.
View return policy
Description
MRP6 Monoclonal specifically detects MRP6 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| MRP6 | |
| Monoclonal | |
| Unconjugated | |
| Tissue culture supernatant with 0.1% Sodium Azide | |
| ABCC6 | |
| Fusion protein containing amino acids 764 to 964 of human MRP6 (FP M6II). | |
| 0.2 mL | |
| Cancer | |
| 368 | |
| Human | |
| Supernatant |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| M6II-31 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Anthracycline resistance-associated protein, ARAPXE, ATP-binding cassette, sub-family C (CFTR/MRP), member 6, EST349056, MLP1ABC34, MOATE, MOAT-E, MRP6ATP-binding cassette sub-family C member 6, multidrug resistance-associated protein 6, Multi-specific organic anion transporter E, pseudoxanthoma elasticum, PXE1, URG7 | |
| Rat | |
| Tissue culture supernatant | |
| RUO | |
| Primary | |
| M6II-31 reacts with MRP6, a 190-200 kD transmembrane protein that is related to the multidrug resistance related protein MRP. Mutations in the MRP6 gene are responsible for the connective tissue disorder Pseudoxanthoma elasticum (PXE). M6II-31 was raised against a bacterial fusion protein of human MRP6, containing amino acids 764-964, spanning the putative 12th transmembrane region as well as predicted internal and external regions of the protein. M6II-7 did not cross-react with the human MDR1, MRP1, MRP2, MRP3, MRP4, MRP5 gene products. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction