missing translation for 'onlineSavingsMsg'
Learn More

MRP6 Antibody (1E6), Novus Biologicals™

Product Code. 18323568 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18323568 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18323568 Supplier Novus Biologicals Supplier No. H00000368M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MRP6 Monoclonal antibody specifically detects MRP6 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRP6
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1E6
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001162
Gene Alias Anthracycline resistance-associated protein, ARAPXE, ATP-binding cassette, sub-family C (CFTR/MRP), member 6, EST349056, MLP1ABC34, MOATE, MOAT-E, MRP6ATP-binding cassette sub-family C member 6, multidrug resistance-associated protein 6, Multi-specific organic anion transporter E, pseudoxanthoma elasticum, PXE1, URG7
Host Species Mouse
Immunogen ABCC6 (NP_001162, 831 a.a. ~ 930 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. IAEMGSYQELLQRKGALVCLLDQARQPGDRGEGETEPGTSTKDPRGTSAGRRPELRRERSIKSVPEKDRTTSEAQTEVPLDDPDRAGWPAGKDSIQYGRV
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 368
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.