missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578695
This item is not returnable.
View return policy
Description
Human ATF1 shares 91% amino acid (aa) sequence identity with mouse ATF1. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - IHC: rat intestine tissue, human intestinal cancer tissue, mouse intestine tissue.
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies. This protein is a member of the MRP subfamily which is involved in multi-drug resistance. The specific function of this protein has not yet been determined; however, this protein may play a role in cellular detoxification as a pump for its substrate, organic anions. Alternative splicing results in multiple splice variants encoding different isoforms.
Specifications
| MRP4 | |
| Polyclonal | |
| Unconjugated | |
| ABCC4 | |
| ABCC4; ABC-tranporter; ATP binding cassette subfamily C member 4; ATP-binding cassette protein C4; ATP-binding cassette sub-family C member 4; ATP-binding cassette, subfamily C (CFTR/MRP), member 4; ATP-binding cassette, sub-family C (CFTR/MRP), member 4; bA464I2.1 (ATP-binding cassette, sub-family C (CFTR/MRP), member 4); canalicular multispecific organic anion transporter (ABC superfamily); D630049P08Rik; EST170205; MOATB; MOAT-B; MRP/cMOAT-related ABC transporter; MRP4; Multidrug resistance-associated protein 4; multi-specific organic anion transporter B; RP11-74A12.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10257, 170924, 239273 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| O15439 | |
| ABCC4 | |
| E.coli-derived human MRP4 recombinant protein (Position: M1-P370). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction