Learn More
MRP4 Mouse anti-Human, Unconjugated, Clone: 1B10B4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67230-1-IG-20UL
This item is not returnable.
View return policy
Description
The MRP4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MRP4. This antibody recognizes human antigen. The MRP4 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| MRP4 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, ELISA | |
| 1B10B4 | |
| Unconjugated | |
| 20 μL | |
| 10257 | |
| IgG1 |