Learn More
Invitrogen™ MRP4 Monoclonal Antibody (M4I-80)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA135682
Description
The M4I-80 Mab was selected after immunization with a fusion protein containing the E. coli maltose binding protein and a fragment of the human MRP4 protein corresponding to amino acids 372-431. MRP4 transports cyclic nucleotides and anti-retroviral compounds. The M4I-80 Mab also reacts with the mouse orthologue of the transporter molecule (Mrp4).
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies. This protein is a member of the MRP subfamily which is involved in multi-drug resistance. The specific function of this protein has not yet been determined; however, this protein may play a role in cellular detoxification as a pump for its substrate, organic anions. Alternative splicing results in multiple splice variants encoding different isoforms.
Specifications
| MRP4 | |
| Monoclonal | |
| 200 μg/mL | |
| tissue culture supernatant with 0.7% BSA and 0.1% sodium azide | |
| O15439 | |
| ABCC4 | |
| MRP4 aa 372-431. | |
| RUO | |
| 10257, 239273 | |
| 4°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry | |
| M4I-80 | |
| Unconjugated | |
| ABCC4 | |
| ABCC4; ABC-tranporter; ATP binding cassette subfamily C member 4; ATP-binding cassette protein C4; ATP-binding cassette sub-family C member 4; ATP-binding cassette, subfamily C (CFTR/MRP), member 4; ATP-binding cassette, sub-family C (CFTR/MRP), member 4; bA464I2.1 (ATP-binding cassette, sub-family C (CFTR/MRP), member 4); canalicular multispecific organic anion transporter (ABC superfamily); D630049P08Rik; EST170205; MOATB; MOAT-B; MRP/cMOAT-related ABC transporter; MRP4; Multidrug resistance-associated protein 4; multi-specific organic anion transporter B; RP11-74A12.1 | |
| Rat | |
| 1 mL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG2a |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.