missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRP4/ABCC4 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
£161.00 - £383.00
Specifications
| Antigen | MRP4/ABCC4 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30229046
|
Novus Biologicals
NBP3-33369-100ul |
100 μL |
£383.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30226620
|
Novus Biologicals
NBP3-33369-20ul |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MRP4/ABCC4 Monoclonal antibody specifically detects MRP4/ABCC4 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| MRP4/ABCC4 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 10257 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ATP-binding cassette sub-family C member 4, ATP-binding cassette, sub-family C (CFTR/MRP), member 4, EST170205, MOAT-BbA464I2.1 (ATP-binding cassette, sub-family C (CFTR/MRP), member 4), MOATBcanalicular multispecific organic anion transporter (ABC superfamily), MRP4MRP/cMOAT-related ABC transporter, multidrug resistance-associated protein 4, multispecific organic anion transporter B, Multi-specific organic anion transporter B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1067-1325 of human MRP4/ABCC4 (NP_005836.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title