missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583912
This item is not returnable.
View return policy
Description
Immunogen sequence: FLPQTDFIIV LADGQVSEMG PYPALLQRNG SFANFLCNYA PDEDQGHLED SWTALEGAED KEALLIEDTL SNHTDLTDND PVTYVVQKQF MRQLSALSSD GEGQGRPVPR RHLGPSEKVQ VTEAKADGAL T Highest antigen sequence indentity to the following orthologs: Mouse - 64%, Rat - 61%.
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MRP subfamily which is involved in multi-drug resistance. The specific function of this protein has not yet been determined; however, this protein may play a role in the transport of biliary and intestinal excretion of organic anions. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
Specifications
| MRP3 | |
| Polyclonal | |
| Unconjugated | |
| ABCC3 | |
| ABC31; ABCC3; ATP binding cassette subfamily C member 3; ATP-binding cassette sub-family C member 3; ATP-binding cassette, subfamily C (CFTR/MRP), member 3; ATP-binding cassette, sub-family C (CFTR/MRP), member 3; canalicular multispecific organic anion transporter 2; canicular multispecific organic anion transporter; cMOAT2; DKFZp686E22157; EST90757; MLP2; MLP-2; MOAT-D; Mrp3; MRP-like protein 2; multidrug resistance associated protein; multidrug resistance protein 3; multidrug resistance-associated protein 3; multi-specific organic anion transporter D; organic anion transporter | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8714 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15438 | |
| ABCC3 | |
| Recombinant protein corresponding to Human ABCC3. Recombinant protein control fragment (Product #RP-103027). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction