missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRP3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13208618
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13208618 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13208618 Supplier Invitrogen™ Supplier No. PA523653

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MRP3 Polyclonal Antibody for Western Blot, IHC (P), Flow

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MRP subfamily which is involved in multi-drug resistance. The specific function of this protein has not yet been determined; however, this protein may play a role in the transport of biliary and intestinal excretion of organic anions. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRP3
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ABCC3
Gene Accession No. O15438
Gene Alias ABC31; ABCC3; ATP binding cassette subfamily C member 3; ATP-binding cassette sub-family C member 3; ATP-binding cassette, subfamily C (CFTR/MRP), member 3; ATP-binding cassette, sub-family C (CFTR/MRP), member 3; canalicular multispecific organic anion transporter 2; canicular multispecific organic anion transporter; cMOAT2; DKFZp686E22157; EST90757; MLP2; MLP-2; MOAT-D; Mrp3; MRP-like protein 2; multidrug resistance associated protein; multidrug resistance protein 3; multidrug resistance-associated protein 3; multi-specific organic anion transporter D; organic anion transporter
Gene Symbols ABCC3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 899-925 amino acids from the central region of human ABCC3.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8714
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.