missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRP3, Mouse anti-Human, Clone: ABCC3/2971, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79703-20UG
This item is not returnable.
View return policy
Description
MRP3 Monoclonal specifically detects MRP3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| MRP3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| O15438 | |
| ABCC3 | |
| Recombinant fragment (around aa 815-957) of human MRP3 protein (exact sequence is proprietary) | |
| 20 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| ABCC3/2971 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| ABC31, ATP-binding cassette sub-family C member 3, ATP-binding cassette, sub-family C (CFTR/MRP), member 3, canalicular multispecific organic anion transporter 2, canicular multispecific organic anion transporter, cMOAT2, EC 3.6.3, EC 3.6.3.44, EC 6.3.2.4, MLP2DKFZp686E22157, MRP3MOAT-DEST90757, multidrug resistance associated protein, Multidrug resistance-associated protein 3, Multi-specific organic anion transporter D | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 8714 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction