missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRP2 Recombinant Rabbit Monoclonal Antibody (23GB4275), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | MRP2 |
|---|---|
| Clone | 23GB4275 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088325
![]() |
Invitrogen™
MA552670 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089006
|
Invitrogen™
MA552671 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MRP2 Recombinant Monoclonal antibody specifically detects MRP2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| MRP2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q92887 | |
| 1244 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4275 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| ABC30; Abcc2; AI173996; ATP binding cassette subfamily C member 2; ATP-binding cassette sub-family C member 2; ATP-binding cassette, subfamily C (CFTR/MRP), member 2; ATP-binding cassette, sub-family C (CFTR/MRP), member 2; calicular multispecific organic anion transporter; canalicular multidrug resistance protein; Canalicular multispecific organic anion transporter 1; Cmoat; CMOAT1; Cmrp; DJS; Human MRP2; KIAA1010; MRP2; multidrug resistance associated protein 2; multidrug resistance protein 2; multidrug resistance-associated protein 2 | |
| A synthesized peptide derived from human MRP2 (1500-1545AA). | |
| Primary | |
| -20°C | |
| ABCC2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title