missing translation for 'onlineSavingsMsg'
Learn More

MRP2 Rabbit anti-Human, Clone: 6L3Z6, Novus Biologicals™

Product Code. 18332116 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18332116 100 μg 100µL
18053984 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18332116 Supplier Novus Biologicals Supplier No. NBP316817100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MRP2 Monoclonal antibody specifically detects MRP2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRP2
Applications Western Blot
Classification Monoclonal
Clone 6L3Z6
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias ATP-binding cassette, sub-family C (CFTR/MRP), member 2, Canalicular multidrug resistance protein, canalicular multispecific organic anion transporter 1, CMOAT1, CMOATABC30, CMRP, DJS, KIAA1010, MRP2ATP-binding cassette sub-family C member 2, Multidrug resistance-associated protein 2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1446-1545 of human MRP2 (Q92887).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline ABC Transporters, Amino Acids Drugs and other small molecules, Cancer, Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1244
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.