missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRP2 Rabbit anti-Human, Clone: 6L3Z6, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | MRP2 |
|---|---|
| Clone | 6L3Z6 |
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18053984
|
Novus Biologicals
NBP3-16817-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18332116
|
Novus Biologicals
NBP3-16817-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MRP2 Monoclonal antibody specifically detects MRP2 in Human samples. It is validated for Western BlotSpecifications
| MRP2 | |
| Western Blot 1:500 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ATP-binding cassette, sub-family C (CFTR/MRP), member 2, Canalicular multidrug resistance protein, canalicular multispecific organic anion transporter 1, CMOAT1, CMOATABC30, CMRP, DJS, KIAA1010, MRP2ATP-binding cassette sub-family C member 2, Multidrug resistance-associated protein 2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1446-1545 of human MRP2 (Q92887). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 6L3Z6 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| ABC Transporters, Amino Acids Drugs and other small molecules, Cancer, Diabetes Research, Lipid and Metabolism | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 1244 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title