missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRP2 Monoclonal Antibody (M2I-4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA126535
This item is not returnable.
View return policy
Description
MA1-26535 detects multidrug resistance associated protein 2 from human samples.
This gene encodes the prototype of the membrane-associated guanylate kinase (MAGUK) family proteins. MAGUKs interact with the cytoskeleton and regulate cell proliferation, signaling pathways, and intercellular junctions. The encoded protein is an extensively palmitoylated membrane phosphoprotein containing a PDZ domain, a Src homology 3 (SH3) motif, and a guanylate kinase domain. This gene product interacts with various cytoskeletal proteins and cell junctional proteins in different tissue and cell types, and may be involved in the regulation of cell shape, hair cell development, neural patterning of the retina, and apico-basal polarity and tumor suppression pathways in non-erythroid cells. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| MRP2 | |
| Monoclonal | |
| Unconjugated | |
| ABCC2 | |
| ABC30; Abcc2; AI173996; ATP binding cassette subfamily C member 2; ATP-binding cassette sub-family C member 2; ATP-binding cassette, subfamily C (CFTR/MRP), member 2; ATP-binding cassette, sub-family C (CFTR/MRP), member 2; calicular multispecific organic anion transporter; canalicular multidrug resistance protein; Canalicular multispecific organic anion transporter 1; Cmoat; CMOAT1; Cmrp; DJS; Human MRP2; KIAA1010; MRP2; multidrug resistance associated protein 2; multidrug resistance protein 2; multidrug resistance-associated protein 2 | |
| Mouse | |
| Purified | |
| RUO | |
| 1244 | |
| 4°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry | |
| M2I-4 | |
| PBS with 1% BSA and 0.1% sodium azide | |
| Q92887 | |
| ABCC2 | |
| Bacterial fusion protein of human MRP 2 containing amino acids of 215-310. | |
| 250 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction